Learn More
IgG(H+L) Donkey anti-Mouse, Biotin-SP, Secondary Antibody, Proteintech
Código de producto. 17690347 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
200 μL
1 mL
Tamaño de la unidad:
1 ml
200 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
17690347 1 mL 1 ml
17600357 200 μL 200 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 17690347 Proveedor Proteintech N.º de proveedor SA0000491ML
Donkey anti-Mouse Affinipure Secondary Antibody
Especificaciones
Antígeno IgG(H+L)
Aplicaciones ELISA, Western Blot, Immunohistochemistry, Immunocytochemistry
Conjugado Biotin-SP
Formulación 0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA
Especie del huésped Donkey
Método de purificación The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads.
Cantidad 1 mL
Primario o secundario Secondary
Especies diana Mouse
Contenido y almacenamiento Store at -20°C. Stable for one year after shipment.