missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG, Goat anti-Rabbit, BV480, Polyclonal, BD Horizon™
Goat Polyclonal Antibody
Marca: BD Biosciences 564879
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
IgG Polyclonal antibody specifically detects IgG in Rabbit samples. It is validated for Bioimaging.
Especificaciones
| IgG | |
| Polyclonal | |
| Brilliant Violet 480 | |
| Goat | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The polyclonal antibody was purified from antiserum by negative adsorption and affinity chromatography. The antibody was conjugated with BD Horizon BV480 under optimum conditions and unconjugated antibody and free BD Horizon BV480 were removed. |
| In vitro Assay | |
| 0.2mg/mL | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| 50 μg | |
| Cell Biology | |
| Rabbit | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Reseach Use Only.
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido