Learn More
IgG, FcγSubclass 2b Specific Goat anti-Mouse, HRP, Secondary Antibody, Proteintech
Código de producto. 17630387 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
1 mL
200 μL
Tamaño de la unidad:
1 ml
200 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
17630387 200 μL 200 microlitros
17620387 1 mL 1 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 17630387 Proveedor Proteintech N.º de proveedor SA000123200UL
Goat anti-Mouse Affinipure Secondary Antibody
Especificaciones
Antígeno IgG, FcγSubclass 2b Specific
Aplicaciones Western Blot
Conjugado HRP
Formulación 0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA
Especie del huésped Goat
Método de purificación The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads.
Cantidad 200 μL
Primario o secundario Secondary
Especies diana Mouse
Contenido y almacenamiento Store at -20°C. Stable for one year after shipment.