missing translation for 'onlineSavingsMsg'
Learn More

PeproTech IGFBP3 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
GREENER_CHOICE

Código de producto. 17895143
Change view
Click to view available options
Cantidad:
2 x 500 μg
25 μg
50 μg
500 μg
Tamaño de la unidad:
1 miligramo
25 microgramos
50 microgramos
500 microgramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17895143 25 μg 25 microgramos
17855713 50 μg 50 microgramos
17895333 2 x 500 μg 1 miligramo
30054436 500 μg 500 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17895143 Proveedor PeproTech N.º de proveedor 500P230BT25UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: GASSGGLGPV VRCEPCDARA LAQCAPPPAV CAELVREPGC GCCLTCALSE GQPCGIYTER CGSGLRCQPS PDEARPLQAL LDGRGLCVNA SAVSRLRAYL LPAPPAPGNA SESEEDRSAG EVESPSVSST HRVSDPKFHP LHSKIIIIKK GHAKDSQRYK VDYESQSTDT QNFSSESKRE TEYGPCRREM EDTLNHLKFL NVLSPRGVHI PNCDKKGFYK KKQCRPSKGR KRGFCWCVDK YGQPLPGYTT KGKEDVHCYS MQSK. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human IGF-BP3. Anti-Human IGF-BP3-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hIGF-BP3 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human IGF-BP3 (500-P230) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hIGF-BP3. Western Blot: To detect hIGF-BP3 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hIGF-BP3 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P230BT-1MG will be provided as 2 x 500 μg

IGFBP3 is a gene that belongs to the insulin-like growth factor binding protein (IGFBP) family. The gene encodes a protein that contains both an IGFBP domain and a thyroglobulin type-I domain. This protein is known to form a ternary complex with insulin-like growth factor acid-labile subunit (IGFALS) and either insulin-like growth factor (IGF) I or II. The complex circulates in the plasma and prolongs the half-life of IGFs while also altering their interaction with cell surface receptors. Various isoforms of the gene have been identified through alternate transcriptional splice variants. Diseases associated with IGFBP3 include Insulin-Like Growth Factor I and Acid-Labile Subunit Deficiency.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno IGFBP3
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Concentración 0.1-1.0 mg/mL
Conjugado Biotin
Formulación PBS with no preservative
génica IGFBP3
N.º de referencia del gen P17936
Alias de gen acid stable subunit of the 140 K IGF complex; AI649005; binding protein 29; binding protein 53; BP53; BP-53; Growth hormone dependent binding protein; growth hormone-dependent binding protein; I79_011450; IBP3; IBP-3; IGF binding protein 3; IGF-binding protein 3; IGFBP; Igfbp3; IGF-BP3; IGFBP-3; IGgfbp3; Insulin like growth factor binding protein; insulin like growth factor binding protein 3; insulin-like growth factor binding protein 3; Insulin-like growth factor binding protein 3 precursor; insulin-like growth factor-binding protein (IGF-BP3); insulin-like growth factor-binding protein 3; tcag7.703
Símbolos de los genes IGFBP3
Especie del huésped Rabbit
Inmunógeno E.coli-derived Recombinant Human IGF-BP3.
Método de purificación Antigen affinity chromatography
Cantidad 25 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3486
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Lyophilized
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.