missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGFBP3 Monoclonal Antibody (OTI3B9), TrueMAB™, OriGene
Descripción
IGFBP3 Monoclonal antibody specifically detects IGFBP3 in Human samples. It is validated for Western Blot
Especificaciones
Especificaciones
| Antígeno | IGFBP3 |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI3B9 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| génica | KCNIP2 |
| N.º de referencia del gen | Q9NS61 |
| Alias de gen | acid stable subunit of the 140 K IGF complex; AI649005; binding protein 29; binding protein 53; BP53; BP-53; Growth hormone dependent binding protein; growth hormone-dependent binding protein; I79_011450; IBP3; IBP-3; IGF binding protein 3; IGF-binding protein 3; IGFBP; Igfbp3; IGF-BP3; IGFBP-3; IGgfbp3; Insulin like growth factor binding protein; insulin like growth factor binding protein 3; insulin-like growth factor binding protein 3; Insulin-like growth factor binding protein 3 precursor; insulin-like growth factor-binding protein (IGF-BP3); insulin-like growth factor-binding protein 3; tcag7.703 |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?