missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IGF2-AS Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA572019
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
IGF2-AS Polyclonal Antibody for Western Blot
The insulin-like growth factors possess growth-promoting activity. In vitro, they are potent mitogens for cultured cells. IGF-II is influenced by placental lactogen and may play a role in fetal development. Preptin undergoes glucose-mediated co-secretion with insulin, and acts as physiological amplifier of glucose-mediated insulin secretion. Exhibits osteogenic properties by increasing osteoblast mitogenic activity through phosphoactivation of MAPK1 and MAPK3.
Especificaciones
| IGF2-AS | |
| Polyclonal | |
| Unconjugated | |
| IGF2-AS | |
| IGF; IGF2 antisense RNA; IGF2AS; IGF2-AS; IGF2-AS1; Insulin like growth factor; PEG8; PEG8/IGF2AS protein | |
| Rabbit | |
| Protein A, Antigen affinity chromatography | |
| RUO | |
| 51214 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.55 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q6U949 | |
| IGF2-AS | |
| KLH conjugated synthetic peptide between 44-73 amino acids from the Central region of human IGF2AS. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido