Learn More
IgE Goat anti-Dog, Polyclonal , Abnova™
Goat polyclonal antibody raised against dog IgE.
Marca: Abnova PAB29765.1mg
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
| IgE | |
| Polyclonal | |
| Goat polyclonal antibody raised against dog IgE. | |
| In PBS, pH 7.2 (0.09% sodium azide) | |
| Dog IgE whole molecule. | |
| 1 mg | |
| Secondary | |
| Dog | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| ELISA (2 ug/mL) Immunoprecipitation (2 ug/mg) Western Blot (0.05-0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| Goat | |
| Antigen Affinity Purified | |
| RUO | |
| The immunoaffinity purified antibodies are specific for dog IgE as determined by immunoelectrophoresis and Ouchterlony double diffusion analysis. | |
| Store at 4°C. | |
| IgG |