missing translation for 'onlineSavingsMsg'
Learn More

IgE Antibody (omalizumab) [Janelia Fluor™ 525], Novus Biologicals™

Product Code. 30167958
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
Product Code. Cantidad unitSize
30167958 0.1 mL 0.10 ml
1 options
This item is not returnable. View return policy

Product Code. 30167958

Brand: Novus Biologicals (Bio-Techne) NBP328478JF525

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Human Monoclonal Antibody

IgE Monoclonal antibody specifically detects IgE in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antígeno IgE
Aplicaciones Flow Cytometry, ELISA, Functional Assay
Clasificación Monoclonal
Clon omalizumab
Conjugado Janelia Fluor 525
Formulación 50mM Sodium Borate
Alias de gen IgE, immunoglobulin heavy constant epsilon
Especie del huésped Human
Inmunógeno IgE
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Adaptive Immunity, Immunology
Primario o secundario Primary
ID de gen (Entrez) 3497
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.