Descripción
The IFITM3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human IFITM3. This antibody recognizes human, mouse, rat antigen. The IFITM3 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™594.
Especificaciones
Especificaciones
| Antígeno | IFITM3 |
| Aplicaciones | Immunofluorescence, Flow Cytometry (Intracellular Staining) |
| Clasificación | Polyclonal |
| Conjugado | CoraLite 594 |
| Especie del huésped | Rabbit |
| Cantidad | 100 μL |
| Primario o secundario | Primary |
| ID de gen (Entrez) | 10410 |
| Especies diana | Human, Rat, Mouse |
| Isotype | IgG |