missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IFI6 Polyclonal Antibody

Descripción
IFI6 Polyclonal Antibody for Western Blot
G1P3, also known as Interferon alpha-inducible protein 6 (IFI6) and Interferon-induced protein 6-16, is a 130 amino acid member of the IFI6 family of proteins. Localized to the mitochondria, G1P3 is a multi-pass membrane protein that is induced by IFN-α2β. G1P3 has been shown to play a major role in the apoptosis pathway. Specifically, G1P3 acts as a cell survival protein by inhibiting caspase-3 activity, which antagonizes apoptosis. G1P3 has been implicated in tumorigenesis and is the subject of therapeutic studies, as reducing G1P3-mediated antiapoptotic signals could suggest improved therapies for myeloma or other malignancies.
Especificaciones
Especificaciones
| Antígeno | IFI6 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.09% sodium azide; pH 7.4 |
| génica | IFI6 |
| N.º de referencia del gen | P09912 |
| Alias de gen | 6-16; FAM14C; G1P3; IFI6; IFI616; IFI-6-16; interferon alpha inducible protein 6; interferon alpha-inducible protein 6; interferon, alpha-inducible protein 6; interferon, alpha-inducible protein clone IFI-6-16; interferon-induced protein 6-16 |
| Símbolos de los genes | IFI6 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?