missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IFI35 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Marca:  Invitrogen™ PA565421

Código de producto. 15918174

  • 578.00€ / 100 microlitros

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

Immunogen sequence: VDVRELLPGSV MLGFARDGVA QRLCQIGQFT VPLGGQQVPL RVSPYVNGEI QKAEIRSQPV PRSVLVLNIP DILDGPELHD VLEIHFQKPT RGGGEVEALT VVPQGQQGLA VFTSESG Highest antigen sequence identity to the following orthologs - mouse 76%, rat 77%.

Interferon-Induced Protein 35 (IFI35), encoded by the IFI35 gene, is located on chromosome 17 and is recognized for its role in immune response modulation, especially under viral stress conditions. Structurally, IFI35 is part of a complex regulatory network that influences non-canonical NF-kB signaling. This process involves the proteasomal processing of p105 into the DNA-binding transcription factor p50, which forms a heterodimer with RELB (RELB/p50) to activate cell chemotaxis autonomously. The gene's dynamic expression under viral stress has been linked to various immune regulatory networks and is crucial for immune cell recruitment and maintenance, particularly M2-like tumor-associated macrophages (TAMs) within the tumor microenvironment. IFI35 upregulation is significant in conditions of neuroinflammation and multiple sclerosis, especially in patients treated with interferons (IFN), indicating its potential as a biomolecular marker. Additionally, IFI35's role in triple-negative breast cancer highlights its impact on antitumor immunity, where its ablation enhances the infiltration of effector CD8+ T cells and sensitizes tumors to anti-PD-1 immunotherapy. This positions IFI35 as a key player in both immune response and cancer treatment strategies.
TRUSTED_SUSTAINABILITY
Especificaciones

Especificaciones

IFI35
Polyclonal
Unconjugated
IFI35
2010008K16Rik; AW986054; IFI35; ifi-35; IFP 35; IFP35; interferon induced protein 35; interferon-induced 35 kDa protein; Interferon-induced 35 kDa protein homolog; interferon-induced protein 35
Rabbit
Antigen affinity chromatography
RUO
3430
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P80217
IFI35
Recombinant Human IFI35. Recombinant protein control fragment (Product #RP-106104).
100 μL
Primary
Human
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Promociones

Promociones

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado