missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IFI16 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA551690
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: KEKFTPKKII AIANYVCRNG FLEVYPFTLV ADVNADRNME IPKGLIRSAS VTPKINQLCS QTKGSFVNGV FEVHKKNVRG EFTYYEIQDN TGKMEVVVHG RLTTINCEEG DKLKLTCFEL APKSGNTGEL RSVIHSHIKV IK Highest antigen sequence identity to the following orthologs: Mouse - 53%, Rat - 39%.
IFI16 (gamma-inteferon-inducible protein 16) binds double-stranded DNA. It binds preferentially to supercoiled DNA and cruciform DNA structures. It seems to be involved in transcriptional regulation and as a transcriptional repressor. IFI16 may have a role in the regulation of hematopoietic differentiation through activation of unknown target genes. It controls cellular proliferation by modulating the functions of cell cycle regulatory factors including p53/TP53 and the retinoblastoma protein. IFI16 is also involved in innate immune response by recognizing viral dsDNA in the cytosol and nucleus. After binding to viral DNA in the cytoplasm, IFI16 recruits TMEM173/STING and mediates the induction of IFN-beta. It is necessary to activate the IFR3 signaling cascade suring human herpes simplex virus 1 infection and promotes the assembly of heterochromatin on herpesviral DNA and inhibiton of viral immediate-early gene expression and replication. IFI16 is also involved in the MTA1-mediated epigenetic regulation of ESR1 expression in breast cancer.
Especificaciones
| IFI16 | |
| Polyclonal | |
| Unconjugated | |
| IFI16 | |
| gamma-interferon-inducible protein 16; IFI 16C; IFI16; Ifi-16; IFNGIP1; interferon gamma inducible protein 16; interferon, gamma-inducible protein 16; interferon-gamma induced protein IFI 16; interferon-inducible myeloid differentiation transcriptional activator; PYHIN2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3428 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q16666 | |
| IFI16 | |
| Recombinant protein corresponding to Human IFI16. Recombinant protein control fragment (Product #RP-101035). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido