Learn More
Invitrogen™ IDO Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA582816
Description
Immunogen sequence: LCSLESNPSV REFVLSKGDA GLREAYDACV KALVSLRSYH LQIVTKYILI PASQQPKENK TSEDPSKLEA KGTGGTDLMN FLK Highest antigen sequence indentity to the following orthologs: Mouse - 48%, Rat - 54%.
IDO1 is an intracellular heme-containing enzyme that catalyzes the oxidative cleavage of the indole ring of several important regulatory molecules like tryptophan, serotonin, and melatonin. By doing this, IDO1 initiates the production of biologically active metabolites, commonly referred to as kynurenines. IDO1 is widely expressed in a variety of human tissues as well as in macrophages and dendritic cells (DCs). In inflammation, interferons (IFNs) act on specific receptors to trigger IDO1 induction. The production of IFN-gamma and induction of IDO1 represent important antimicrobial mechanisms. Degradation and depletion of tryptophan by IDO1 inhibits the growth of viruses, bacteria and parasites. Furthermore, IDO1 plays a complex and crucial role in immunoregulation during infection, pregnancy, autoimmunity, transplantation, and neoplasia.
Spécification
| IDO | |
| Polyclonal | |
| Unconjugated | |
| Ido1 | |
| EC 1.13.11.52; IDO; IDO1; IDO-1; INDO; indolamine 2,3 dioxygenase; indole 2,3-dioxygenase; indoleamine; indoleamine 2,3-dioxygenase 1; indoleamine 23-dioxygenase; Indoleamine-2; indoleamine-pyrrole 2,3 dioxygenase; indoleamine-pyrrole 2,3-dioxygenase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3620 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P14902 | |
| Ido1 | |
| Recombinant protein corresponding to Human IDO1. Recombinant protein control fragment (Product #RP-93785). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.