Learn More
STEMCELL Technologies IDE1
Descripción
- Inducer of definitive endoderm 1 (IDE1) induces differentiation of mouse or human pluripotent stem cells (PSCs) by activating SMAD2 phosphorylation and NODAL expression (Borowiak et al.) At EC50 = 125 nM, SOX17 expression was induced in mouse ES cells
- Induces differentiation of mouse or human ES cells to definitive endoderm in the absence of Activin A, NODAL, or feeder cells (Borowiak et al.)
Especificaciones
Especificaciones
| CAS | 1160927-48-9 |
| Cantidad | 1 mg |
| Fórmula molecular | C15H18N2O5 |
| Pureza | ≥95% |