Learn More
Invitrogen™ ICAM-3 Monoclonal Antibody (ICAM3.1)
GREENER_CHOICE
Código de producto. 11551062
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
11551062 100 μg 100 microgramos
1 options
Código de producto. 11551062
Marca: Invitrogen™ MA190977
Mouse Monoclonal Antibody
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CD50 (intracellular adhesion molecule 3, ICAM-3) is a transmembrane glycoprotein expressed by leukocytes, that serves as a counter-receptor for the lymphocyte function-associated antigen (LFA)-1 integrin. Besides functioning as an adhesive molecule that mediates e.g. the contact between T cells and antigen presenting cells, ICAM-3 regulates affinity of LFA-1 for ICAM-1 and induces T cell activation and proliferation. ICAM-3 plays an essential role in the initiation of the immune response both on T cells and antigen presenting cells and interacts also with CD209 (dendritic cell-specific ICAM-3-grabbing nonintegrin, DC-SIGN), a C-type lectin of dendritic cells and macrophages; this process is involved in dialogue between dendritic cells and granulocytes.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno ICAM-3
Aplicaciones ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Radioimmune Assays (RIA), Western Blot
Clasificación Monoclonal
Clon ICAM3.1
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with no preservative
génica ICAM3
N.º de referencia del gen P32942
Alias de gen CD50; CDw50; ICAM3; ICAM-3; ICAM-R; Intercellular adhesion molecule 3; intercellular adhesion molecule-3
Símbolos de los genes ICAM3
Especie del huésped Mouse
Inmunógeno ICAM 3Fc chimeric fusion protein.
Método de purificación Protein A
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3385
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1