Learn More
Invitrogen™ iBind™ Flex Multi-Strip Insert Replacement
Descripción
The iBind™ Flex Multi-Strip Insert Replacement is an accessory for the iBind™ Flex Western Device (Cat. No. SLF2000) to process up to six vertically cut strip blots.
iBind Flex Western System
The iBind™ Flex Western System offers automated western blot processing and can accommodate various blot formats including midi, mini, and vertically cut strips. Based on Sequential Lateral Flow (SLF) technology, all blocking, antibody incubation and washing steps during immunodetection are hands-free. Compared to manual methods, the iBind Flex Western System requires less primary antibody, enables reproducible results and is compatible with downstream detection protocols.
Especificaciones
Especificaciones
| Tipo de producto | Inserto de pocillos multitira; con seis pocillos en las filas 1 y 3 |
| Descripción | Multi-strip well insert with six wells on row 1 and 3 |
| Contenido del kit | Seis pocillos en las filas 1 y 3. |
| Para utilizar con (aplicación) | Inmunotransferencia (Western Blotting) |
| Para utilizar con (equipo) | iBind™ |
| Línea de productos | iBind |
| Tipo | Inserto de repuesto |
| Cantidad | 1 inserto |
| Condiciones de envío | Temperatura ambiente |