Learn More
Brady™ i75 Series R4400 Ribbon for the i7500 Label Printer
Código de producto. p-200109826
Click to view available options
Color:
Azul
Blanco
Rojo
Verde
Anchura (métrico):
60 mm
83 mm
110 mm
Tamaño de la unidad:
cada uno
Código de producto. 30279184
Marca: Brady™ 176777
Solo quedan null
Añadir a la cesta
Añadir a la cesta
i75-Series ribbon with LabelSense™ Technology warns user if ribbon is incompatible with installed i75-Series label material.
  • i75-series ribbon is self-centering with ink-side always correctly facing the label
  • One-color ribbon for printing a single color onto a label
  • R4400 Series High-Density Colored Resin formulation offers excellent resistance to solvents, scratches and high heat when printed on the appropriate label material from Brady™
  • Recommended for printing on glossy or very smooth label stocks, such as B-483 Ultra Aggressive Polyester and B-423 Permanent Polyester from Brady™
  • Combine Brady label materials with their recommended ribbon series for superior legend performance
Especificaciones
Tipo de producto Printer Ribbon
Color Blanco
Para utilizar con (equipo) i7500
Material Polyester
Print Technology Ink Jet
Longitud (imperial) 328.08 yd.
Anchura (imperial) 2.36 in.
Longitud (métrico) 300 m
Anchura (métrico) 60 mm
Grosor (métrico) 4.5 μm
Dimensiones (L x An) 60 mm x 300 m
Cantidad 1 Ea.
Alternate Part Number i75-R4400-WT