Learn More
Brady™ i75 Series R4400 Ribbon for the i7500 Label Printer
Descripción
- i75-series ribbon is self-centering with ink-side always correctly facing the label
- One-color ribbon for printing a single color onto a label
- R4400 Series High-Density Colored Resin formulation offers excellent resistance to solvents, scratches and high heat when printed on the appropriate label material from Brady™
- Recommended for printing on glossy or very smooth label stocks, such as B-483 Ultra Aggressive Polyester and B-423 Permanent Polyester from Brady™
- Combine Brady label materials with their recommended ribbon series for superior legend performance
Especificaciones
Especificaciones
| Tipo de producto | Printer Ribbon |
| Color | Rojo |
| Para utilizar con (equipo) | i7500 |
| Material | Polyester |
| Print Technology | Ink Jet |
| Longitud (imperial) | 328.08 yd. |
| Anchura (imperial) | 2.36 in. |
| Longitud (métrico) | 300 m |
| Anchura (métrico) | 60 mm |
| Grosor (métrico) | 4.5 μm |
| Mostrar más |