Learn More
STEMCELL Technologies HypoThermosol™ FRS
Código de producto. 17139921
Encompass_Preferred
Change view
Click to view available options
Cantidad:
100 mL
16 x 10 mL
Bolsa de 500 mL
Frasco de 500 mL
Tamaño de la unidad:
100 ml
500 ml
paquete de 16
17139921 16 x 10 mL paquete de 16
17109931 Bolsa de 500 mL 500 ml
17149921 100 mL 100 ml
17159921 Frasco de 500 mL 500 ml
4 options
Código de producto. 17139921
Marca: STEMCELL Technologies 07934
Animal component-free, defined hypothermic (2 - 8°C) preservation medium for a range of cell and tissue types
  • HypoThermosol™ FRS is an optimized hypothermic (2 - 8°C) preservation medium that enables improved and extended preservation of cells, tissues, and organs
  • HypoThermosol™ FRS is uniquely formulated to address the molecular-biological response of cells during the hypothermic preservation process
  • HypoThermosol™ FRS includes key ions at concentrations that balance the intracellular state at hypothermic temperatures
  • Additional components include pH buffers, energy substrates, free radical scavengers, and osmotic/oncotic stabilizers
Especificaciones
Tipo de célula Hematopoietic Stem and Progenitor Cells; Mesenchymal Stem and Progenitor Cells; Other; Pluripotent Stem Cells
Para utilizar con (aplicación) Crioconservación
Cantidad 16 x 10 mL