Learn More
Thermo Scientific™ HyperSep™ Servo+ Total-B SPE Cartridges and Well Plates
Código de producto. 11372293 Tienda Thermo Scientific Productos
Change view
Click to view available options
Descripción:
Columna para SPE de 100 mg 6 mL, paquete de 30
Columna para SPE de 150 mg 6 mL, paquete de 30
Columna para SPE de 1 g 25 mL, paquete de 20
Columna para SPE de 200 mg 6 mL, paquete de 30
Columna para SPE de 30 mg/1 mL, paquete de 100
Columna para SPE de 500 mg 6 mL, paquete de 30
Columna para SPE de 500 mg/3 mL, paquete de 50
Columna para SPE de 60 mg 6 mL, paquete de 30
Placa de 96 pocillos, 10 mg, 1 unidad
Placa de 96 pocillos, 5 mg, 1 unidad
Peso del lecho:
5 mg
10 mg
30 mg
60 mg
100 mg
150 mg
200 mg
500 mg
1 g
Volumen (métrico):
1 mL
3 mL
6 mL
25 mL
Tamaño de la unidad:
cada uno
paquete de 100
paquete de 20
paquete de 30
paquete de 50
Código de producto. Descripción Peso del lecho Volumen (métrico) unitSize
11372293 Columna para SPE de 150 mg 6 mL, paquete de 30 150 mg 6 mL paquete de 30
11352203 Columna para SPE de 30 mg/1 mL, paquete de 100 30 mg 1 mL paquete de 100
11362203 Placa de 96 pocillos, 5 mg, 1 unidad 5 mg - cada uno
11382203 Placa de 96 pocillos, 10 mg, 1 unidad 10 mg - cada uno
11332293 Columna para SPE de 500 mg/3 mL, paquete de 50 500 mg 3 mL paquete de 50
11342293 Columna para SPE de 500 mg 6 mL, paquete de 30 500 mg 6 mL paquete de 30
11352293 Columna para SPE de 100 mg 6 mL, paquete de 30 100 mg 6 mL paquete de 30
11362293 Columna para SPE de 60 mg 6 mL, paquete de 30 60 mg 6 mL paquete de 30
11382293 Columna para SPE de 200 mg 6 mL, paquete de 30 200 mg 6 mL paquete de 30
11392293 Columna para SPE de 1 g 25 mL, paquete de 20 1 g 25 mL paquete de 20
10 options
Código de producto. 11372293
Marca: Thermo Scientific™ 60110309C
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos
Screening for unknown basic compounds when analyzing drugs of abuse? These SPE cartridges and plates offer a robust, ready-to-use solution.
Get the cleanest possible sample extracts—and maximum confidence—when preparing samples for screening for drugs of abuse. Depending on your analytical needs, you can choose from a comprehensive range of Thermo Scientific™ HyperSep Servo+ products, each optimized for a different class of drugs. To ensure success, each product comes with detailed extraction procedures.
Thermo Scientific™ HyperSep Servo+ Total-B Cartridges and Well Plates offer a robust, ready-to-use solution for screening of a wide range of unknown basic compounds.
HyperSep Servo and Servo+ Cartridges and Plates
Thermo Scientific™ HyperSep Servo and Servo+ cartridges and plates provide complete, high-reliability SPE sample preparation solutions for drug screening applications.
Both Servo and Servo+ product lines deliver the high levels of reproducibility and clean sample extracts required for maximum confidence when dealing with complex biological sample matrices. Servo+ cartridges and plates offer an extra measure of selectivity, higher loading capacities and enhanced robustness.
HyperSep SPE Cartridges and Plates
Thermo Scientific™ HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.
TRUSTED_SUSTAINABILITY
Especificaciones
Descripción Columna para SPE de 150 mg 6 mL, paquete de 30
Peso del lecho 150 mg
Para utilizar con (aplicación) Analysis of Drugs of Abuse
Línea de productos HyperSep
Absorbente HyperSep Servo+ Total B
Tipo Columnas de extracción en fase sólida
Volumen (métrico) 6 mL