Learn More
Thermo Scientific™ HyPerforma™ Smartainer™ 3.0 Bioprocess Container (BPC), Aegis™ 5-14 film
GREENER_CHOICE
Código de producto. 17948221 Tienda Thermo Scientific Productos
Change view
Click to view available options
Capacidad (métrico):
200 L
500 L
Tipo de inserto:
Con fuelle horizontal
Con fuelle vertical
Tamaño de la unidad:
paquete de 5
Código de producto. Capacidad (métrico) Tipo de inserto unitSize
17948221 500 L Con fuelle horizontal paquete de 5
17958221 200 L Con fuelle horizontal paquete de 5
17978261 500 L Con fuelle vertical paquete de 5
3 options
Código de producto. 17948221
Marca: Thermo Scientific™ SH31188.02
The Thermo Scientific HyPerforma Smartainer 3.0 BPC provides end-users producing biologics or vaccine therapeutics with a large-volume, sterile liquid storage system for volumes ranging from 200 L to 3000 L—designed for the collection and storage of sterile process liquids such as cell culture media, process water or purification media. Options include temperature-control through integrated water jackets, BPC deploying hoists, as well as other options to optimize performance in your productions processes.
The Thermo Scientific HyPerforma Smartainer 3.0 BPC provides end-users producing biologics or vaccine therapeutics with a large-volume, sterile liquid storage system for volumes ranging from 200 L to 3000 L—designed for the collection and storage of sterile process liquids such as cell culture media, process water or purification media. Options include temperature-control through integrated water jackets, BPC deploying hoists, as well as other options to optimize performance in your productions processes.
TRUSTED_SUSTAINABILITY
Especificaciones
Capacidad (métrico) 500 L
Product Type Bioprocess Container (BPC)
Tipo de inserto Con fuelle horizontal