Learn More
STEMCELL Technologies Hydrocortisone Stock Solution
Descripción
- Hydrocortisone Stock Solution is recommended for use as a cell culture supplement and for the preparation of PneumaCult™, MammoCult™, and EpiCult™ media when culturing airway epithelial cells, mammary primary cells, or tumor cell lines
- Hydrocortisone Stock Solution is provided at a concentration of 96 μg/mL, which is ideal as a 200X stock solution for use with PneumaCult™-ALI (Catalog #05001), MammoCult™ (Catalog #05620), EpiCult™-B (Human; Catalog #05601), and EpiCult™-C (Catalog #05630) media
- It may also be used for preparing PneumaCult™-Ex Medium (Catalog #05008) or PneumaCult™-Ex Plus Medium (Catalog #05040)
Especificaciones
Especificaciones
| Tipo de célula | Airway Cells, Cancer Cells and Cell Lines, Mammary Cells |
| Para utilizar con (aplicación) | Cultivo celular |
| Cantidad | 3 mL |