Learn More
Juchheim Laborgeraete™ Gato hidráulico para laboratorio
Código de producto. p-7229368
Change view
Click to view available options
Altura (métrico):
625 mm
1060 mm
Tamaño de la unidad:
cada uno
Código de producto. Altura (métrico) unitSize
10294772 625 mm cada uno
10204782 1060 mm cada uno
2 options
Código de producto. 10294772
Marca: Juchheim Laborgeraete™ 02294
Solo quedan null
Añadir a la cesta
Añadir a la cesta
A laboratory lifting table with smooth-running lifting hydraulics that loads up to 85 kg.
  • For static and dynamic loads
  • Particularly stable due to scissor construction made of robust steel
  • Acid and base resistant due to high-quality powder coating
  • Requires no external connections (pump hydraulics fully integrated)
  • With handles for easy transport
  • To lower the load, the lever only has to be pressed slightly upwards. The load then sinks slowly (approx. 3 cm/s) and stops as soon as the lever is released again.
Especificaciones
Material Acero
Altura (métrico) 625 mm
Anchura (métrico) 400 mm
Descripción Adjustment Range: 175 to 625 mm
Peso (métrico) 30 kg