Learn More
GE Healthcare Hyclone™ HyClone™ Irradiated Super Low IgG Fetal Bovine Serum
Código de producto. 12370143
Change view
Click to view available options
Cantidad:
500 mL
Tamaño de la unidad:
500 ml
12370143 500 mL 500 ml
1 options
Código de producto. 12370143
Marca: GE Healthcare Hyclone™ SH30898.03IR
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos
Processed to substantially reduce IgG (via Protein G chromatography) level, HyClone super low IgG fetal bovine serum (FBS) is suitable for use in more exacting applications.
  • FBS contains natural levels of immunoglobulin (IgG) that may be too high for certain cell culture and protein purification applications.
  • Serum from individual animals may be screened for low content and pooled to produce large batches of FBS with lower levels of IgG. However, this may be quite a challenge due to the scarcity of such low IgG animals and the logistics involved with the screening and collection process.
  • Even naturally occurring low levels of IgG may still be too high for certain applications.
  • Comparative levels of IgG in bovine serum products are shown in the table below. 
  • HyClone® Super Low IgG FBS performance is equivalent to non-processed FBS and is suitable for applications requiring lower levels of specific antibody, such as propagation of bovine viruses and monoclonal antibody production.
TRUSTED_SUSTAINABILITY
Especificaciones
Contenido y almacenamiento –10°C
País de origen Estados Unidos
Tipo de producto Fetal Bovine Serum
Cantidad 500 mL