Learn More
HVCN1 Rabbit anti-Human, Polyclonal, Proteintech
Código de producto. 16882674 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16882674 150 μL 150 microlitros
16892674 20 μL 20 microlitros
2 options
Código de producto. 16882674
Marca: Proteintech 141621AP150UL
Rabbit Polyclonal Antibody
HVCN1 (hydrogen voltage-gated channel 1), a voltage gated proton channel protein that is highly expressed in immune tissues, mediates the proton conductances required by phagocytic leukocytes for the oxidative burst that underlies microbial killing. HVCN1 moderates the voltage-dependent proton permeability of excitable membranes by allowing the flow of protons in accordance to their electrochemical gradient. HVCN1 is sensitive to zinc ions, and can be inhibited by them. HVCN1 may represent a new therapeutic target for B cell maligcies on continued signaling via the B cell antigen receptor.
Especificaciones
Antígeno HVCN1
Aplicaciones Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Clasificación Polyclonal
Concentración 0.3 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica HVCN1
N.º de referencia del gen Q96D96
Alias de gen HV1, HVCN1, VSOP
Símbolos de los genes HVCN1
Especie del huésped Rabbit
Inmunógeno HVCN1 Fusion Protein Ag5350
Método de purificación Antigen Affinity Chromatography
Cantidad 150 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 84329
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG