missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The protein encoded by this gene is a component of an evolutionarily conserved, genotoxin-activated checkpoint complex that is involved in the cell cycle arrest in response to DNA damage. This protein forms a heterotrimeric complex with checkpoint proteins RAD9 and RAD1.
Specifications
Specifications
| Antígeno | HUS1 |
| Aplicaciones | Western Blot, Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Concentración | 0.3 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| génica | HUS1 |
| N.º de referencia del gen | O60921 |
| Alias de gen | Checkpoint protein HUS1, hHUS1, HUS1 |
| Símbolos de los genes | HUS1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?