missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ZO-1 ELISA Kit
Descripción
ZO-1 forms complexes with either ZO-2 or ZO-3. In addition, these proteins can also associate with claudin, occludin and F-actin, at tight junction stands, where they provide a linkage between the actin cytoskeleton and the tight junction. ZO-1 expression is significantly reduced in many breast cancer lines. ZO-2 and ZO-3 are ubiquitously expressed within epithelial tight junctions, and unlike ZO-1, which is also expressed at cell junctions of cardiac myocytes, ZO-2 is not expressed in nonepithelial tissue. ZO-1 is a protein located on a cytoplasmic membrane surface of intercellular tight junctions. ZO-1 may be involved in signal transduction at cell-cell junctions. Two transcript variants encoding distinct isoforms have been identified for the ZO-1 gene. Diseases associated with ZO-1 dysfunction include Celiac Disease and Congenital Nephrotic Syndrome.
Especificaciones
Especificaciones
| Número de acceso | Q07157 |
| Intervalo del ensayo | 0.16 to 10 ng/mL |
| Sensibilidad del ensayo | 0.09 ng/mL |
| Conjugado | HRP |
| Tipo de producto | ELISA |
| Tipo de muestra | Plasma, Serum |
| Para utilizar con (equipo) | Colorimetric Microplate Reader |
| ID de gen (Entrez) | 7082 |
| Símbolo de gen | TJP1 |
| CV interensayo | <10 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?