missing translation for 'onlineSavingsMsg'
Learn More

Human VCAM-1/CD106 Antibody, R&D Systems™
GREENER_CHOICE

Código de producto. 18722608
Change view
Click to view available options
Cantidad:
100 μg
25 μg
500 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
500 microgramos
Código de producto. Cantidad unitSize
18722608 25 μg 25 microgramos
18788478 100 μg 100 microgramos
18749986 500 μg 500 microgramos
3 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18722608

Marca: R&D Systems MAB809SP

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody has been used in 4 publications

VCAM-1/CD106 Monoclonal specifically detects VCAM-1/CD106 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY

Specifications

Antígeno VCAM-1/CD106
Aplicaciones ELISA
Clasificación Monoclonal
Clon HAE-2Z
Conjugado Unconjugated
Dilución ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Alias de gen CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1
Símbolos de los genes VCAM1
Especie del huésped Mouse
Inmunógeno Human VCAM-1
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 25 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 7412
Especificidad de la prueba Detects recombinant human VCAM-1 in direct ELISAs and Western blots.
Reconstitución Reconstitute at 0.5 mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Formulario Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.