missing translation for 'onlineSavingsMsg'
Learn More

Human u-Plasminogen Activator (uPA)/ Urokinase Catalytic Domain Antibody, R&D Systems™
GREENER_CHOICE

Código de producto. 18712438
Change view
Click to view available options
Cantidad:
100 μg
25 μg
500 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
500 microgramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18712438 500 μg 500 microgramos
18756399 25 μg 25 microgramos
18779428 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18712438 Proveedor R&D Systems N.º de proveedor MAB1310500

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody has been used in 3 publications

u-Plasminogen Activator/Urokinase Monoclonal specifically detects u-Plasminogen Activator/Urokinase in Human samples. It is validated for Western Blot, Immunoprecipitation, ELISA Capture (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno u-Plasminogen Activator/Urokinase
Aplicaciones Western Blot, Immunoprecipitation, ELISA
Clasificación Monoclonal
Clon 204212
Concentración LYOPH
Conjugado Unconjugated
Dilución Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
N.º de referencia del gen P00749
Alias de gen ATF, EC 3.4.21, EC 3.4.21.73, plasminogen activator, urokinase, PLAU, uPA, u-PA, uPlasminogen Activator, U-plasminogen activator, urinary, Urokinase, urokinase-type plasminogen activator
Símbolos de los genes PLAU
Especie del huésped Mouse
Inmunógeno Mouse myeloma cell line NS0-derived recombinant human u-Plasminogen Activator (uPA)/Urokinase Catalytic Domain Ser21-Leu431 Accession # P00749
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 500 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 5328
Especificidad de la prueba Detects human u-Plasminogen Activator (uPA)/Urokinase Catalytic Domain in direct ELISAs and Western blots. In Western blots under reducing conditions, it reacts with the catalytic domain (B chain) only. In Western blots under non-reducing conditions, it reacts with the B chain and disulfide bond-linked B and A chains. In direct ELISAs, no cross-reactivity with recombinant human (rh) Factor X, rhHGFA, rhKLK-3, -4, -5, -6, -8, -10, -11, rhThrombin, or recombinant mouse WIF-1 is observed.
Reconstitución Reconstitute at 0.5 mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Formulario Supernatant
Isotype IgG2a
Mostrar más Mostrar menos

For Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.