missing translation for 'onlineSavingsMsg'
Learn More

Human TSC1 Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Código de producto. 18725449
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18725449 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18725449 Proveedor R&D Systems N.º de proveedor IC4379T100UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

TSC1 Monoclonal specifically detects TSC1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno TSC1
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon 488915
Concentración 0.2 mg/mL
Conjugado Alexa Fluor 594
Dilución Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulación Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
N.º de referencia del gen Q92574
Alias de gen Hamartin, KIAA0243MGC86987, LAMhamartin, TSC, tuberous sclerosis 1, Tuberous sclerosis 1 protein, tumor suppressor
Símbolos de los genes TSC1
Especie del huésped Mouse
Inmunógeno E. coli-derived recombinant human TSC1 Asp156-Thr300 Accession # Q92574
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 7248
Especificidad de la prueba Detects human TSC1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) TSC2 (aa 550-850), rhTSC2 (aa 1506-1748), or recombinant mouse TSC22 (aa 1-143) is observed.
Especies diana Human
Contenido y almacenamiento Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.