Learn More
Human TREM2 Antibody, R&D Systems™
GREENER_CHOICE
Código de producto. 18781117
Change view
Click to view available options
Cantidad:
100 μg
25 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
18781117 25 μg 25 microgramos
18743667 100 μg 100 microgramos
2 options
Código de producto. 18781117
Marca: R&D Systems MAB18282SP
Mouse Monoclonal Antibody
TREM2 Monoclonal antibody specifically detects TREM2 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno TREM2
Aplicaciones Immunohistochemistry
Clasificación Monoclonal
Clon 1057114
Conjugado Unconjugated
Dilución Immunohistochemistry 5-25 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
Alias de gen PLOSL2, TREM-2, Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2
Especie del huésped Mouse
Inmunógeno NS0-derived recombiHumant human TREM-2 protein, His19-Ser174
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 25 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 54209
Reconstitución Reconstitute at 0.5 mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Formulario Purified
Isotype IgG1