Learn More
Novus Biologicals™ Human TNF-alpha ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Marca: Novus Biologicals™ NBP1-91170
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The Human TNF-alpha ELISA Kit (Colorimetric) measures human TNF-alpha in cell culture supernatant, serum, plasma (edta, citrate, heparin).Especificaciones
| 7.8 - 500pg/mL | |
| Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum | |
| ELISA | |
| TNF | |
| Adaptive Immunity, Asthma, Autophagy, Biologically Active Proteins, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity | |
| APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necr |
| 2.3pg/mL | |
| Human | |
| 7124 | |
| 96 Tests | |
| Storage of components varies. See protocol for specific instructions. | |
| TNF-alpha |