Learn More
Human TAPBPR Antibody, Novus Biologicals™
GREENER_CHOICE
Código de producto. 18776125
Change view
Click to view available options
Cantidad:
100 μg
25 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
18776125 100 μg 100 microgramos
18795965 25 μg 25 microgramos
2 options
Código de producto. 18776125
Marca: R&D Systems MAB11337100
Mouse Monoclonal Antibody
Tapasin Related Protein Monoclonal antibody specifically detects Tapasin Related Protein in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Tapasin Related Protein
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon 1059329
Conjugado Unconjugated
Dilución Flow Cytometry 0.25 ug/10^6 cells
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
N.º de referencia del gen Q9BX59.2
Alias de gen FLJ10143, TAP binding protein-like, Tapasin-like, TAPASIN-R, tapasin-related protein, TAP-binding protein-like, TAP-binding protein-related protein, TAPBPL, TAPBPR, TAPBP-RTAP binding protein related, TAPBPRtapasin-like
Especie del huésped Mouse
Inmunógeno Chinese Hamster Ovary cell line, CHO-derived human TAPBPR, Ala19-Arg404, Accession # Q9BX59.2
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 55080
Reconstitución Reconstitute at 0.5 mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Formulario Purified
Isotype IgG2a