Learn More
Human Syndecan-2/CD362 Antibody, R&D Systems™
GREENER_CHOICE
Código de producto. 18763378
Change view
Click to view available options
Cantidad:
1 mg
100 μg
25 μg
500 ug
Tamaño de la unidad:
1 miligramo
100 microgramos
25 microgramos
500 microgramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
18763378 25 μg 25 microgramos
18729458 100 μg 100 microgramos
18149640 1 mg 1 miligramo
18171661 500 ug 500 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 18763378 Proveedor R&D Systems N.º de proveedor MAB2965RSP
Rat Monoclonal Antibody
Syndecan-2/CD362 Monoclonal specifically detects Syndecan-2/CD362 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Syndecan-2/CD362
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon 305515R
Conjugado Unconjugated
Dilución Flow Cytometry 0.25 ug/10^6 cells
Formulación Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
N.º de referencia del gen AAH49836
Alias de gen CD362, CD362 antigen, Fibroglycan, heparan sulfate proteoglycan 1, cell surface-associated, Heparan sulfate proteoglycan core protein, HSPG, HSPG1, HSPG1syndecan proteoglycan 2, SDC2, SYND2, SYND2cell surface-associated heparan sulfate proteoglycan 1, syndecan 2, syndecan-2, Syndecan2
Símbolos de los genes SDC2
Especie del huésped Rat
Inmunógeno Mouse myeloma cell line NS0-derived recombinant human Syndecan-2/CD362 Glu19-Gly144 Accession # AAH49836
Método de purificación Protein A or G purified from cell culture supernatant
Cantidad 25 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 6383
Especificidad de la prueba Detects human Syndecan-2/CD362 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Syndecan-1, recombinant human (rh) Syndecan-3, or rhSyndecan-4 is observed.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Formulario Purified
Isotype IgG2b
Mostrar más Mostrar menos
For Research Use Only