missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human STING/TMEM173 Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 5 publications
Marca: R&D Systems AF6516
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
STING/TMEM173 Polyclonal specifically detects STING/TMEM173 in Human samples. It is validated for Western Blot, Simple Western, Knockout Validated.
Especificaciones
| STING/TMEM173 | |
| Polyclonal | |
| Unconjugated | |
| Q86WV6 | |
| TMEM173 | |
| E. coli-derived recombinant human STING/TMEM173 Ala215-Ser379 Accession # Q86WV6 | |
| 100 μg | |
| Primary | |
| Detects human STING/TMEM173 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| endoplasmic reticulum IFN stimulator, Endoplasmic reticulum interferon stimulator, ERIS, FLJ38577, hMITA, Mediator of IRF3 activation, MITA, mitochondrial mediator of IRF3 activation, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner, Stimulator of interferon genes protein, sting, STINGhSTING, TMEM173, transmembrane protein 173 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 340061 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido