Learn More
Human SR-BI Antibody, R&D Systems™
GREENER_CHOICE
Código de producto. 18792628
Change view
Click to view available options
Cantidad:
100 μg
25 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18792628 25 μg 25 microgramos
18192661 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18792628 Proveedor R&D Systems N.º de proveedor MAB8114SP
Mouse Monoclonal Antibody
SR-BI Monoclonal specifically detects SR-BI in Human samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno SR-BI
Aplicaciones Western Blot, Immunohistochemistry
Clasificación Monoclonal
Clon 947007
Conjugado Unconjugated
Dilución Western Blot 2 ug/mL, Immunohistochemistry 5-25 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Alias de gen CD36 and LIMPII analogous 1, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1, CD36L1, CD36L1scavenger receptor class B type 1, CLA1, CLA1 CD36 antigen-like 1, CLA-1 HDLQTL6, Collagen type I receptor, thrombospondin receptor-like 1, HDLQTL6, MGC138242, SCARB1, scavenger receptor class B type III, scavenger receptor class B, member 1, SR-B1, SRB1 scavenger receptor class B member 1, SRBI
Símbolos de los genes SCARB1
Especie del huésped Mouse
Inmunógeno Human embryonic kidney cell line HEK293-derived human SR-BI
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 25 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 949
Especificidad de la prueba Detects human SR-BI in direct ELISAs and Western blots.
Reconstitución Reconstitute at 0.5 mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Formulario Purified
Isotype IgG1
Mostrar más Mostrar menos
For Research Use Only