Learn More
Invitrogen™ Human SP-D ELISA Kit
Human SP-D quantitates human SP-D in serum, plasma, supernatant.
Marca: Invitrogen™ EH436RBX10
3041.80 EUR hasta el 2023-03-27
Use el código promocional "20585" para aplicar la oferta.
Detalles adicionales : Peso : 2.50000kg
Descripción
Human SP-D quantitates human SP-D in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human SP-D.
Surfactant protein D (SP-D) is synthesized and secreted by lung epithelial cells. It belongs to group III of the family of C-type lectins and members of this group has overall structure consisting of multiple globular 'head' regions linked by triple-helical, collagen-like, strands. This group also includes SP-A and the serum proteins mannan-binding protein, conglutinin and collectin-43, all of which have been shown to bind to the C1q receptor found on a wide variety of cells. Both SP-D and SP-A have been shown to enhance oxygen radical production by alveolar macrophages.Especificaciones
AI573415; COLEC7; Collectin7; Collectin-7; CP4; EGK_19692; Lung surfactant protein D; PSPD; PSP-D; pulmonary surfactant apoprotein; pulmonary surfactant protein D; pulmonary surfactant-associated protein D; Sftp4; SFTPD; SPD; SP-D; surfactant associated protein D; surfactant protein D; surfactant, pulmonary-associated protein D; surfactant-associated protein, pulmonary 4 | |
0.12 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
SFTPD | |
<10% | |
HRP | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |
0.12 to 30 ng/mL | |
Biotin | |
Supernatant | |
P35247 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
10 x 96 Tests | |
Serum, 50 μL; Supernatant, 50 μL; Plasma, 50 μL | |
SFTPD | |
4 hr 45 min |