missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SLITRK1 Synthetic Peptide
Descripción
Slitrk1 Synthetic Peptide, PEP-0590, from Invitrogen.
SLIT and NTRK-like family 1 (Slitrk1) is a member a protein family consisting of six homologous transmembrane proteins (Slitrk1-6) that share two conserved leucine-rich repeat domains in the extracellular domain and have significant homology to Slit, a secreted axonal growth-controlling protein. These proteins are also homologous to trk neurotrophin receptors in their intracellular domains. Expression of Slitrk proteins is highly restricted to neural and brain tumor tissues, but varies within the family. For example, Slitrk1 is expressed primarily in mature neurons. Overexpression of Slitrk1 in transfected neuronal cells induced unipolar neurites, while expression of the other Slitrk proteins inhibited neurite outgrowth, suggesting that these proteins are involved in the control of neurite outgrowth. While Slitrk1 variants have been suggested associated with Tourette's Syndrome, it is thought to play only a minor role if at all.
Especificaciones
Especificaciones
| Gene ID (Entrez) | 114798 |
| Especie | Human |
| Método de purificación | purified |
| Número de acceso | Q96PX8 |
| Nombre común | Human SLITRK1 |
| Concentración | 200 μg/mL |
| Contenido y almacenamiento | -20°C |
| Descripción | Synthetic Peptide |
| Formato | Liquid |
| Formulación | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?