missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SLC22A1 Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems FAB6469T-100UG
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
SLC22A1 Monoclonal antibody specifically detects SLC22A1 in Human samples. It is validated for Flow Cytometry
Especificaciones
| SLC22A1 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| hOCT1, OCT1, oct1_cds, OCT1solute carrier family 22 member 1, Organic cation transporter 1, solute carrier family 22 (organic cation transporter), member 1 | |
| NS0 mouse myeloma cell line transfected with human SLC22A1, Met1-Thr554, Accession # NP_003048 | |
| 100 μg | |
| Primary | |
| Reconstitution default not known | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG2a |
| Flow Cytometry | |
| 609222 | |
| Flow Cytometry 0.25-1 ug/106 cells | |
| NP_003048 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6580 | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido