Learn More
Human SLAM/CD150 Antibody, R&D Systems™
GREENER_CHOICE
Código de producto. 18761207
Change view
Click to view available options
Cantidad:
100 μg
25 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
18761207 100 μg 100 microgramos
18730087 25 μg 25 microgramos
2 options
Código de producto. 18761207
Marca: R&D Systems MAB16421100
Mouse Monoclonal Antibody
SLAM/CD150 Monoclonal specifically detects SLAM/CD150 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno SLAM/CD150
Aplicaciones ELISA
Clasificación Monoclonal
Clon 123303
Conjugado Unconjugated
Dilución ELISA
Formulación Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
N.º de referencia del gen Q13291
Alias de gen CD 150, CD150, CD150IPO-3, CDw150, IPO-3, signaling lymphocytic activation molecule, signaling lymphocytic activation molecule family member 1, SLAMCD150 antigen, SLAMF1
Especie del huésped Mouse
Inmunógeno Mouse myeloma cell line NS0-derived recombinant human SLAM/CD150, Ala21-Pro237, Accession # Q13291
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 6504
Especificidad de la prueba Detects human SLAM/CD150 in direct ELISAs.
Reconstitución Reconstitute at 0.5 mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Formulario Purified
Isotype IgG1