missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Siglec-14 (NP_001092082) VersaClone cDNA
Click to view available options
Cantidad:
10 μg
Tamaño de la unidad:
10 microgramos
Descripción
This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.
The Human Siglec-14 (NP_001092082) VersaClone cDNA clone contains the complete ORF for Siglec-14, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Siglec-14 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Información de órdenes
Ships at ambient temperature
Especificaciones
Especificaciones
| génica | SIGLEC14 |
| Número de acceso | NP_001092082 |
| Concentración | 10 μg at 0.2 μg/μL |
| Formulación | cDNA is provided in 10mM Tris-Cl, pH 8.266 |
| Tipo de inserto | 1204bp |
| ID de gen (Entrez) | 100049587 |
| Alias de gen | AC018755.2, sialic acid binding Ig-like lectin 14, Siglec14 |
| Contenido y almacenamiento | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Especie | Human |
| Cantidad | 10 μg |
For Reasearch Use Only. Not for use in Humans.
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido