missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Serum Albumin Monoclonal Antibody (AL-01), Biotin

Description
This antibody reacts only with human serum albumin, a 65-67 kDa monomeric protein in human blood plasma; it is produced in liver. No cross-reactivity was observed with other serum proteins.
Albumin is a soluble, monomeric protein with a molecular weight of approximately 65-67 kDa that comprises about one-half of the blood serum protein. Human serum albumin (HAS) is the most abundant protein in human blood plasma, and has a serum half-life of approximately 20 days. Albumin functions primarily as a carrier protein for steroids, fatty acids, and thyroid hormones and plays a role in stabilizing extracellular fluid volume. Albumin is a globular, unglycosylated serum protein synthesized in the liver as pre-pro-albumin which has an N-terminal peptide that is removed before the nascent protein is released from the rough produce the secreted albumin. Serum albumin has a good binding capacity for water, Ca(2+), Na(+), K(+), fatty acids, hormones, bilirubin and drugs. The main function of albumin is the regulation of the colloidal osmotic pressure of blood.
Specifications
Specifications
| Antígeno | Human Serum Albumin |
| Aplicaciones | ELISA, Immunohistochemistry (Paraffin), Radioimmune Assays (RIA), Western Blot |
| Clasificación | Monoclonal |
| Clon | AL-01 |
| Concentración | 1 mg/mL |
| Conjugado | Biotin |
| Formulación | PBS with 15mM sodium azide; pH 7.4 |
| génica | ALB |
| N.º de referencia del gen | P02768 |
| Alias de gen | Alb; Alb1; Alb-1; albumin; albumin (32 AA); albumin (AA 34); albumin 1; Albza; ANALBA; cell growth inhibiting protein 42; DKFZp779N1935; FDAH; GIG20; GIG42; growth-inhibiting protein 20; PRO0883; PRO0903; PRO1341; PRO1708; PRO2044; PRO2619; PRO2675; Serum albumin; UNQ696/PRO1341 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?