missing translation for 'onlineSavingsMsg'
Learn More

Human Serpin E1/PAI-1 Antibody, R&D Systems™
GREENER_CHOICE

Código de producto. 18709287
Change view
Click to view available options
Cantidad:
25 μg
Tamaño de la unidad:
25 microgramos
Código de producto. Cantidad unitSize
18709287 25 μg 25 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18709287

Marca: R&D Systems AF1786SP

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Goat Polyclonal Antibody has been used in 4 publications

Serpin E1/PAI-1 Polyclonal specifically detects Serpin E1/PAI-1 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunoprecipitation, Neutralization.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Serpin E1/PAI-1
Aplicaciones Western Blot, Immunohistochemistry, Immunoprecipitation, Neutralization
Clasificación Polyclonal
Concentración LYOPH
Conjugado Unconjugated
Dilución Western Blot 1 ug/mL, Simple Western 2.5 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 3.5 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
N.º de referencia del gen P05121
Alias de gen Endothelial plasminogen activator inhibitor, Nexin, PAI1, PAI-1, PAI1PAI-1, PAISerpin E1, PLANH1, PLANH1plasminogen activator inhibitor 1, serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 1, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 1
Símbolos de los genes SERPINE1
Especie del huésped Goat
Inmunógeno S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Serpin E1/PAI-1 Gly21-Pro402 Accession # P05121
Método de purificación Affinity Purified
Cantidad 25 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 5054
Especificidad de la prueba Detects human Serpin E1/PAI-1 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse Serpin E1 is observed and less than 2% cross-reactivity with recombinant human (rh) Serpin A1, rhSerpin A3, rhSerpin A4, rhSerpin A5, rhSerpin C1, rhSerpin F2, and rhSerpin F1/PEDF is observed.
Reconstitución Reconstitute at 0.2 mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Mostrar más Mostrar menos

For Research Use Only

Título del producto
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.