Learn More
Invitrogen™ Human Serpin A3/Alpha-1-Antichymotrypsin ELISA Kit
Human Serpin A3/Alpha-1-Antichymotrypsin quantitates human Serpin A3/Alpha-1-Antichymotrypsin in serum, plasma, supernatant.
Marca: Invitrogen™ EH411RBX10
Detalles adicionales : Peso : 2.50000kg
Descripción
Human Serpin A3/Alpha-1-Antichymotrypsin quantitates human Serpin A3/Alpha-1-Antichymotrypsin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Serpin A3/Alpha-1-Antichymotrypsin.
SERPINA encodes Alpha 1-ACT. It is an early-stage acute-phase plasma protein and a serpin that preferentially inactivates chymotrypsin, cathepsin G, and chymase. Alpha-1-ACT, a serine protease inhibitor, is tightly associated with amyloid plaques in Alzheimer's disease (AD) and in normal aged human and monkey brain. Regulation of the serine proteases and serine protease inhibitors plays an important role in neuromuscular differentiation. Prostate specific antigen (PSA), a chymotrypsin-like serine protease, is predominantly complexed to Alpha-1-ACT.Especificaciones
AACT; ACT; Alpha-1-antichymotrypsin; Alpha-1-antichymotrypsin His-Pro-less; Cell growth-inhibiting gene 24/25 protein; GIG24; GIG25; growth-inhibiting protein 24; growth-inhibiting protein 25; MGC88254; serine (or cysteine) proteinase inhibitor, clade A, member 3; serpin A3; serpin family A member 3; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 3; SERPINA3 | |
0.14 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
SERPINA3 | |
<10% | |
HRP | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |
0.137 to 100 ng/mL | |
Biotin | |
Supernatant | |
P01011 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
10 x 96 Tests | |
Serum, 50 μL; Supernatant, 50 μL; Plasma, 50 μL | |
SERPINA3 | |
4 hr 45 min |