Learn More
R&D Systems™ Human SCF Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Marca: R&D Systems™ 255-SC-01M
Includes: For Research Use Only
Descripción
Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. Kitamura, T. et al. (1989) J. Cell Physiol. 140:323. The ED50 for this effect is 1-5 ng/mL. The specific activity of Recombinant Human SCF is approximately 1.3 x 106 IU/mg, which is calibrated against recombinant human SCF WHO International Standard (NIBSC code: 91/682). Specific activity is for reference purposes only and is not routinely tested.Información de órdenes
Ships at ambient temperature.
Especificaciones
| P21583.1 | |
| 4254 | |
| 1mg | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
| <0.10 EU per 1μg of the protein by the LAL method | |
| 1 to 5ng/mL | |
| Unconjugated |
| Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein | |
| M.W. (theroretical): 19 kDa | |
| E. coli-derived human SCF/c-kit Ligand protein Glu26-Ala189,with an N-terminal Met | |
| RUO | |
| c-kit Ligand, DCUA, DFNA69, DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, FPHH, KIT ligand, Kitl, KITLG, KL-1, Mast cell growth factor, MGF, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, SHEP7, SLF, steel factor | |
| Recombinant protein | |
| Yes |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
For Research Use Only