missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Rat LRP-4 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Marca: R&D Systems AF5948-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
LRP-4 Polyclonal specifically detects LRP-4 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Especificaciones
| LRP-4 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CLSS, KIAA0816, low density lipoprotein receptor-related protein 4, low-density lipoprotein receptor-related protein 4, LRP10, LRP4, LRP-4, MEGF7LRP10, Multiple epidermal growth factor-like domains 7 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 4038 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| O75096 | |
| LRP4 | |
| E. coli-derived recombinant human LRP-4 Gly18-Ser344 Accession # O75096 | |
| 25 μg | |
| Primary | |
| Detects human and rat LRP-4 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) LRP-1, rhLRP-1B, rhLRP-5, rhLRP-6, and rhLDL R is observed. | |
| Human, Rat | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido