missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Proprotein Convertase 9/PCSK9 Biotinylated Antibody, R&D Systems™
Sheep Polyclonal Antibody
Marca: R&D Systems BAF3888
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Proprotein Convertase 9/PCSK9 Polyclonal specifically detects Proprotein Convertase 9/PCSK9 in Human samples. It is validated for Western Blot.
Especificaciones
| Proprotein Convertase 9/PCSK9 | |
| Polyclonal | |
| Biotin | |
| Q8NBP7 | |
| PCSK9 | |
| Mouse myeloma cell line NS0-derived recombinant human Proprotein Convertase 9/PCSK9 Gln31-Gln692 Accession # Q8NBP7 | |
| 50 μg | |
| Primary | |
| Detects human Proprotein Convertase 9/PCSK9 in Western blots. In Western blots, approximately 30% cross-reactivity with recombinant mouse PCSK9 is observed and less than 1% cross-reactivity with recombinant human (rh) PCSK1 and rhPCSK7 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| EC 3.4.21, EC 3.4.21.111, FH3, FH3neural apoptosis regulated convertase 1, FHCL3, HCHOLA3, hypercholesterolemia, autosomal dominant 3, LDLCQ1, NARC1, NARC-1, NARC-1convertase subtilisin/kexin type 9 preproprotein, NARC1EC 3.4.21.-, Neural apoptosis-regulated convertase 1, PC9, PCSK9, Proprotein convertase 9, proprotein convertase subtilisin/kexin type 9, Subtilisin/kexin-like protease PC9 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 255738 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido