missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Pro-BMP-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Marca: R&D Systems MAB2260-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
BMP-2 Monoclonal specifically detects BMP-2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Especificaciones
| BMP-2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BDA2, BMP2, BMP-2, BMP-2A, BMP2ABone morphogenetic protein 2A, bone morphogenetic protein 2, SSFSC | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 650 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| 253717 | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| P12643 | |
| BMP2 | |
| Mouse myeloma cell line NS0-derived recombinant human BMP-2 Ala22-Lys281 (Arg245Ser, Arg279Ser) Accession # P12643 | |
| 25 μg | |
| Primary | |
| Detects the pro region of the human BMP-2 precursor in direct ELISAs and Western blots. This antibody does not recognize mature recombinant human BMP-2. | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido