missing translation for 'onlineSavingsMsg'
Learn More

Human PKC epsilon Antibody, R&D Systems™
GREENER_CHOICE

Código de producto. 18179960
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
Código de producto. Cantidad unitSize
18179960 100 μg 100 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18179960

Marca: R&D Systems MAB5134

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

PKC epsilon Monoclonal specifically detects PKC epsilon in Human samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno PKC epsilon
Aplicaciones Immunohistochemistry
Clasificación Monoclonal
Clon 666823
Concentración LYOPH
Conjugado Unconjugated
Dilución Immunohistochemistry 8-25 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
N.º de referencia del gen Q02156
Alias de gen EC 2.7.11, EC 2.7.11.13, MGC125656, nPKC-epsilon, PKCE, PKCEMGC125657, PRKCE, protein kinase C epsilon type, protein kinase C, epsilon
Símbolos de los genes PRKCE
Especie del huésped Mouse
Inmunógeno E. coli-derived recombinant human PKC epsilon Gln580-Pro737 Accession # Q02156
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 5581
Especificidad de la prueba Detects human PKC epsilon in direct ELISAs. In direct ELISAs, less than 20% cross-reactivity with recombinant human (rh) PKC delta and rhPKC iota is observed. No cross-reactivity with rhPKC alpha , beta 1, beta 2, gamma , eta , μ, nu , theta , or zeta is observed.
Reconstitución Sterile PBS to a final concentration of 0.5 mg/mL.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Formulario Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.