Learn More
Human Phospho-STAT3 (Y705) Antibody, R&D Systems™
Código de producto. 18101811
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
18101811 100 μg 100 microgramos
1 options
Código de producto. 18101811
Marca: R&D Systems MAB4607
Rat Monoclonal Antibody has been used in 2 publications
STAT3 Monoclonal specifically detects STAT3 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno STAT3 (p tyr705)
Aplicaciones Western Blot
Clasificación Monoclonal
Clon 805435
Concentración LYOPH
Conjugado Unconjugated
Dilución Western Blot 2 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Alias de gen Acute-phase response factor, APRFMGC16063, DNA-binding protein APRF, FLJ20882, HIES, signal transducer and activator of transcription 3, signal transducer and activator of transcription 3 (acute-phase responsefactor)
Símbolos de los genes STAT3
Especie del huésped Rat
Inmunógeno Phosphopeptide containing the human STAT3 Y705 site
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 6774
Especificidad de la prueba Detects human STAT3 when phosphorylated at Y705.
Reconstitución Sterile PBS to a final concentration of 0.5 mg/mL.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Formulario Purified
Isotype IgG2b